Assay Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148123.1 | NP_001141595.1 | 1704 | 73 | ||
| GenBank mRNA | BT041686.1 | - | 1704 | 73 | ||
| BT063423.1 | - | 1375 | 73 | |||
More Information
Set Membership: |
|

CER1
Genomic Map