Assay Details
Gene Symbol: LOC100280076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153016.1 | NP_001146488.1 | 875 | 86 | ||
GenBank mRNA | BT055522.1 | - | 875 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
