Assay Details
Gene Symbol: LOC100280629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153547.1 | NP_001147019.1 | 1320 | 101 | ||
GenBank mRNA | EU952967.1 | - | 1320 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
