Assay Details
Gene Symbol: LOC100282811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155717.1 | NP_001149189.1 | 433 | 95 | ||
GenBank mRNA | EU962381.1 | - | 433 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
