Assay Details
Gene Symbol: LOC100284394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157289.1 | NP_001150761.1 | 1139 | 68 | ||
GenBank mRNA | BT068309.1 | - | 1112 | 68 | ||
EU968142.1 | - | 1139 | 68 |
Gene Symbol: LOC100501579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT085532.1 | - | 1225 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
