Assay Details
Gene Symbol: LOC100275291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001149410.1 | NP_001142882.1 | 486 | 97 | ||
GenBank mRNA | BT068910.1 | - | 486 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
