Assay Details
| Gene Symbol: LOC100280482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153402.1 | NP_001146874.1 | 753 | 89 | ||
| GenBank mRNA | EU951986.1 | - | 753 | 89 | ||
More Information
Set Membership: |
|

LOC100280482
Genomic Map