Assay Details
Gene Symbol: LOC100280970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153890.1 | NP_001147362.1 | 287 | 94 | ||
GenBank mRNA | EU954969.1 | - | 287 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
