Assay Details
| Gene Symbol: LOC100282186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155098.1 | NP_001148570.1 | 241 | 75 | ||
| GenBank mRNA | BT068502.1 | - | 127 | 75 | ||
| EU959964.1 | - | 241 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282186| Gene Symbol: LOC100282186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155098.1 | NP_001148570.1 | 241 | 75 | ||
| GenBank mRNA | BT068502.1 | - | 127 | 75 | ||
| EU959964.1 | - | 241 | 75 | |||