Assay Details
Gene Symbol: LOC100283232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156134.1 | NP_001149606.1 | 331 | 65 | ||
GenBank mRNA | EU963937.1 | - | 331 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
