Assay Details
Gene Symbol: LOC100283743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156642.1 | NP_001150114.1 | 1138 | 62 | ||
GenBank mRNA | BT041934.2 | - | 1074 | 62 | ||
EU965769.1 | - | 1138 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
