Assay Details
| Gene Symbol: LOC100284783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157678.1 | NP_001151150.1 | 899 | 85 | ||
| GenBank mRNA | BT065971.1 | - | 899 | 85 | ||
| EU969670.1 | - | 839 | 85 | |||
More Information
Set Membership: |
|

LOC100284783
Genomic Map