Assay Details
Gene Symbol: LOC100284798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157693.1 | NP_001151165.1 | 1050 | 72 | ||
GenBank mRNA | EU969715.1 | - | 1050 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
