Assay Details
Gene Symbol: IDP1992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157734.1 | NP_001151206.1 | 475 | 84 | ||
GenBank mRNA | BT088178.1 | - | 436 | 84 | ||
EU907936.1 | - | 258 | 84 | |||
EU969855.1 | - | 475 | 84 |
