Assay Details
Gene Symbol: LOC100284854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157749.1 | NP_001151221.1 | 819 | 105 | ||
GenBank mRNA | BT066380.1 | - | 1350 | 105 | ||
BT069368.1 | - | 822 | 105 | |||
EU969912.1 | - | 819 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
