Assay Details
Gene Symbol: CNR01 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157851.1 | NP_001151323.1 | 333 | 69 | ||
GenBank mRNA | EU970260.1 | - | 333 | 69 | ||
HM008653.1 | - | 266 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
