Assay Details
Gene Symbol: AC225718.2_FG006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158005.1 | NP_001151477.1 | 251 | 72 | ||
GenBank mRNA | EU970876.1 | - | 251 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
