Assay Details
| Gene Symbol: LOC100285540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158431.1 | NP_001151903.1 | 509 | 64 | ||
| GenBank mRNA | EU972753.1 | - | 509 | 64 | ||
More Information
Set Membership: |
|

LOC100285540
Genomic Map