Assay Details
Gene Symbol: LOC100285540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158431.1 | NP_001151903.1 | 790 | 96 | ||
GenBank mRNA | EU972753.1 | - | 790 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
