Assay Details
| Gene Symbol: LOC100272842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147295.1 | NP_001140767.1 | 973 | 97 | ||
| GenBank mRNA | BT039577.1 | - | 973 | 97 | ||
| BT041030.1 | - | 1127 | 97 | |||
More Information
Set Membership: |
|

LOC100272842
Genomic Map