Assay Details
| Gene Symbol: prl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147950.1 | NP_001141422.1 | 2012 | 65 | ||
| GenBank mRNA | BT041345.1 | - | 2012 | 65 | ||
| BT063632.1 | - | 2012 | 65 | |||
| BT069295.1 | - | 1979 | 65 | |||
More Information
Set Membership: |
|

prl1
Genomic Map