Assay Details
Gene Symbol: prl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147950.1 | NP_001141422.1 | 1200 | 87 | ||
GenBank mRNA | BT041345.1 | - | 1200 | 87 | ||
BT063632.1 | - | 1200 | 87 | |||
BT069295.1 | - | 1167 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
