Assay Details
| Gene Symbol: gpm517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152483.1 | NP_001145955.1 | 271 | 97 | ||
| GenBank mRNA | BT054321.1 | - | 271 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gpm517| Gene Symbol: gpm517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152483.1 | NP_001145955.1 | 271 | 97 | ||
| GenBank mRNA | BT054321.1 | - | 271 | 97 | ||
Set Membership: |
|