Assay Details
Gene Symbol: gpm517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152483.1 | NP_001145955.1 | 357 | 76 | ||
GenBank mRNA | BT054321.1 | - | 357 | 76 | ||
BT061586.1 | - | 485 | 76 |
Gene Symbol: LOC100501645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001196325.1 | NP_001183254.1 | 522 | 76 | ||
GenBank mRNA | BT085858.1 | - | 522 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT085858.1, NM_001196325.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
