Assay Details
| Gene Symbol: LOC100280500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153420.1 | NP_001146892.1 | 1804 | 101 | ||
| GenBank mRNA | EU952136.1 | - | 1804 | 101 | ||
More Information
Set Membership: |
|

LOC100280500
Genomic Map