Assay Details
Gene Symbol: LOC100280673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153592.1 | NP_001147064.1 | 1340 | 108 | ||
GenBank mRNA | EU953205.1 | - | 1340 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
