Assay Details
| Gene Symbol: LOC100280848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153768.1 | NP_001147240.1 | 668 | 76 | ||
| GenBank mRNA | BT039090.1 | - | 497 | 76 | ||
| EU953565.1 | - | 523 | 76 | |||
| EU954192.1 | - | 668 | 76 | |||
More Information
Set Membership: |
|

LOC100280848
Genomic Map