Assay Details
Gene Symbol: LOC100280967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153887.1 | NP_001147359.1 | 753 | 60 | ||
GenBank mRNA | EU954962.1 | - | 753 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
