Assay Details
Gene Symbol: pco069505(751) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154177.1 | NP_001147649.1 | 534 | 90 | ||
GenBank mRNA | BT067057.1 | - | 859 | 90 | ||
EU676033.1 | - | 356 | 90 | |||
EU956135.1 | - | 534 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
