Assay Details
Gene Symbol: pco069505(751) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154177.1 | NP_001147649.1 | 928 | 67 | ||
GenBank mRNA | BT067057.1 | - | 1253 | 67 | ||
EU956135.1 | - | 928 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
