Assay Details
Gene Symbol: LOC100281326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154244.1 | NP_001147716.1 | 979 | 83 | ||
GenBank mRNA | EU956349.1 | - | 979 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
