Assay Details
Gene Symbol: LOC100282570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155478.1 | NP_001148950.1 | 1598 | 73 | ||
GenBank mRNA | EU961494.1 | - | 1598 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
