Assay Details
| Gene Symbol: fus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156244.1 | NP_001149716.1 | 1262 | 77 | ||
| GenBank mRNA | EU964381.1 | - | 1262 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

fus6| Gene Symbol: fus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156244.1 | NP_001149716.1 | 1262 | 77 | ||
| GenBank mRNA | EU964381.1 | - | 1262 | 77 | ||