Assay Details
| Gene Symbol: LOC100283803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156702.1 | NP_001150174.1 | 620 | 79 | ||
| GenBank mRNA | EU966004.1 | - | 620 | 79 | ||
| EU968170.1 | - | 544 | 79 | |||
More Information
Set Membership: |
|

LOC100283803
Genomic Map