Assay Details
Gene Symbol: LOC100284642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157537.1 | NP_001151009.1 | 555 | 84 | ||
GenBank mRNA | EU969156.1 | - | 555 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
