Assay Details
Gene Symbol: LOC100284648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157543.1 | NP_001151015.1 | 427 | 63 | ||
GenBank mRNA | BT085847.1 | - | 543 | 63 | ||
EU969175.1 | - | 427 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
