Assay Details
Gene Symbol: LOC100285133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158027.1 | NP_001151499.1 | 1228 | 69 | ||
GenBank mRNA | EU970973.1 | - | 1228 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
