Assay Details
Gene Symbol: LOC100285438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158331.1 | NP_001151803.1 | 700 | 73 | ||
GenBank mRNA | EU972237.1 | - | 700 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
