Assay Details
| Gene Symbol: LOC100286216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159104.1 | NP_001152576.1 | 394 | 76 | ||
| GenBank mRNA | EU976187.1 | - | 394 | 76 | ||
More Information
Set Membership: |
|

LOC100286216
Genomic Map