Assay Details
| Gene Symbol: rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111420.1 | NP_001104890.1 | 450 | 94 | ||
| GenBank mRNA | AF034948.1 | - | 450 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl17| Gene Symbol: rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111420.1 | NP_001104890.1 | 450 | 94 | ||
| GenBank mRNA | AF034948.1 | - | 450 | 94 | ||
Set Membership: |
|