Assay Details
| Gene Symbol: rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111449.1 | NP_001104919.1 | 961 | 70 | ||
| GenBank mRNA | AF079429.1 | - | 961 | 70 | ||
| BT061958.1 | - | 954 | 70 | |||
More Information
Set Membership: |
|

rad51b
Genomic Map