Assay Details
Gene Symbol: sus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111724.1 | NP_001105194.1 | 2130 | 58 | ||
GenBank mRNA | AY104856.1 | - | 1973 | 58 | ||
AY124703.1 | - | 2130 | 58 |
