Assay Details
| Gene Symbol: inra1(tmp) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111996.1 | NP_001105466.1 | 683 | 72 | ||
| GenBank mRNA | X82633.1 | - | 683 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

inra1(tmp)| Gene Symbol: inra1(tmp) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111996.1 | NP_001105466.1 | 683 | 72 | ||
| GenBank mRNA | X82633.1 | - | 683 | 72 | ||
Set Membership: |
|