Assay Details
| Gene Symbol: pdlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112138.1 | NP_001105608.1 | 899 | 79 | ||
| GenBank mRNA | AF038586.1 | - | 899 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdlk2| Gene Symbol: pdlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112138.1 | NP_001105608.1 | 899 | 79 | ||
| GenBank mRNA | AF038586.1 | - | 899 | 79 | ||
Set Membership: |
|