Assay Details
| Gene Symbol: zds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112139.1 | NP_001105609.1 | 1195 | 86 | ||
| GenBank mRNA | AF047490.1 | - | 1195 | 86 | ||
| AY110101.1 | - | 1195 | 86 | |||
More Information
Set Membership: |
|

zds1
Genomic Map