Assay Details
| Gene Symbol: pdi8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112334.1 | NP_001105804.1 | 1197 | 86 | ||
| GenBank mRNA | AY739290.1 | - | 1197 | 86 | ||
| BT033408.1 | - | 1200 | 86 | |||
More Information
Set Membership: |
|

pdi8
Genomic Map