Assay Details
Gene Symbol: LON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112425.1 | NP_001105895.1 | 1526 | 66 | ||
GenBank mRNA | U85495.1 | - | 1526 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
