Assay Details
| Gene Symbol: LON2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112425.1 | NP_001105895.1 | 2253 | 65 | ||
| GenBank mRNA | U85495.1 | - | 2253 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LON2| Gene Symbol: LON2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112425.1 | NP_001105895.1 | 2253 | 65 | ||
| GenBank mRNA | U85495.1 | - | 2253 | 65 | ||
Set Membership: |
|