Assay Details
| Gene Symbol: apt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112596.1 | NP_001106066.1 | 6460 | 126 | ||
| GenBank mRNA | DQ020098.1 | - | 6460 | 126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

apt1| Gene Symbol: apt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112596.1 | NP_001106066.1 | 6460 | 126 | ||
| GenBank mRNA | DQ020098.1 | - | 6460 | 126 | ||