Assay Details
| Gene Symbol: LOC100126542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112597.1 | NP_001106067.1 | 2295 | 93 | ||
| GenBank mRNA | EF051578.1 | - | 2295 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100126542| Gene Symbol: LOC100126542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112597.1 | NP_001106067.1 | 2295 | 93 | ||
| GenBank mRNA | EF051578.1 | - | 2295 | 93 | ||